Learn More
DUSP19 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA549858
Catalog No. PA549858
Add to cart
Description
Description
The antibody detects endogenous levels of total DUSP19 protein.
Dual-specificity phosphatases (DUSPs) constitute a large heterogeneous subgroup of the type I cysteine-based protein-tyrosine phosphatase superfamily. DUSPs are characterized by their ability to dephosphorylate both tyrosine and serine/threonine residues. They have been implicated as major modulators of critical signaling pathways. DUSP19 contains a variation of the consensus DUSP C-terminal catalytic domain, with the last serine residue replaced by alanine, and lacks the N-terminal CH2 domain found in the MKP (mitogen-activated protein kinase phosphatase) class of DUSPs (see MIM 600714) (summary by Patterson et al., 2009 [PubMed 19228121]).
Specifications
Specifications
DUSP19
Polyclonal
Unconjugated
DUSP19
Liquid
DUSP19
Synthesized peptide derived from internal of human DUS19.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8WTR2
DUSP17, LMWDSP3, SKRP1
Rabbit
IgG
100 μL
-20°C
Primary
142679