Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DUSP23 Mouse anti-Human, Mouse, Rat, APC, Clone: OTI3C10, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271976APC
Description
DUSP23 Monoclonal specifically detects DUSP23 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
DUSP23 | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
DUSP23 | |
Full length recombinant protein of human DUSP23 (NP_060293) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Flow Cytometry | |
OTI3C10 | |
Flow Cytometry | |
dual specificity phosphatase 23, dual specificity protein phosphatase 23, DUSP25, EC 3.1.3.16, EC 3.1.3.48, FLJ20442, LDP3, LDP-3, Low molecular mass dual specificity phosphatase 3, low-molecular-mass dual-specificity phosphatase 3, MOSP, RP11-190A12.1, VH1-like member Z, VH1-like phosphatase Z, VHZ | |
Mouse | |
Affinity Purified | |
GPCR, Protein Phosphatase | |
54935.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title