Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUSP23 Mouse anti-Human, Clone: OTI3C10, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525782

Catalog No. PIMA525782


Add to cart

Description

Description

Myelin / Oligodendrocyte Specific Protein (MOSP) (pI of ~6.7) is found exclusively on oligodendrocytes and myelin in the central nervous system (CNS). Myelin is produced by oligodendrocytes in the CNS and Schwann cells in the peripheral nervous system (PNS). Defining the differences between CNS and PNS myelin may provide insights into the process of myelination and the pathogenesis of certain idiopathic diseases involving myelin such as multiple sclerosis, certain leukodystrophies, and other myelinopathies. On demyelination, immunoreactivity for MOSP, like MBP, is lost.
Specifications

Specifications

DUSP23
Monoclonal
0.94 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BVJ7
DUSP25; LDP-3; MOSP; RP11-190A12.1; VHZ; VH1-like member Z; VH1-like phosphatase Z; dual specificity protein phosphatase 23; low molecular mass dual specificity phosphatase 3; low-molecular-mass dual-specificity phosphatase 3
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54935
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI3C10
Unconjugated
DUSP23
Liquid
DUSP23
Full length human recombit protein of DUSP23 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit