Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUSP4 Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI7C11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270610AF405

Catalog No. N270610A405


Add to cart

Description

Description

DUSP4 Monoclonal antibody specifically detects DUSP4 in Human samples. It is validated for Western Blot.
Specifications

Specifications

DUSP4
OTI7C11
Western Blot
Affinity Purified, Conjugated
DUSP4
Recombinant protein fragment corresponding to amino acids 1-257 of human DUSP4 (NP_476499) produced in E.coli.
Immunogen affinity purified
Cell Cycle and Replication
Primary
1846.0
Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
dual specificity phosphatase 4, Dual specificity protein phosphatase hVH2, EC 3.1.3.16, EC 3.1.3.48, HVH2serine/threonine specific protein phosphatase, MAP kinase phosphatase 2, Mitogen-activated protein kinase phosphatase 2, MKP-2dual specificity protein phosphatase 4, MKP2VH1 homologous phosphatase 2, TYP, VH2
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit