Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DUSP5 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179716
Description
DUSP5 Polyclonal specifically detects DUSP5 in Human samples. It is validated for Western Blot.Specifications
DUSP5 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
NP_004410 | |
DUSP5 | |
Synthetic peptide directed towards the middle region of human DUSP5. Peptide sequence: AGSSLIGHLQTLSPDMQGAYCTFPASVLAPVPTHSTVSELSRSPVATATS | |
Affinity Purified | |
RUO | |
Primary | |
Predicted Homology Based On Immunogen Sequence: Mouse: 86%; Rabbit: 86%; Bovine: 85%; Rat: 79%; Guinea pig: 79%. | |
Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
dual specificity phosphatase 5, dual specificity protein phosphatase 5, Dual specificity protein phosphatase hVH3, DUSP, EC 3.1.3.16, EC 3.1.3.48, HVH3VH1-like phosphatase 3, serine/threonine specific protein phosphatase, VH3 | |
Rabbit | |
42 kDa | |
100 μL | |
Protein Phosphatase | |
1847 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title