Learn More
Bio-Serv™ Dustless Precision Pellets™ Grain-Based Rodent Diet
Precise size and weight pellets generally used in nutritional and behavioral studies
Manufacturer: Bio-Serv™ F0172
Description
- Nutritionally complete grain-based formula
- Dustless and fracture free—superior performance in automatic feeders
- Every batch tested for hardness, weight and height
- Nutritionally assayed
Ingredients: Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Magnesium Stearate, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin
Specifications
3.35 | |
Rodent Diet | |
Nutritional and behavioral studies | |
0.75 g | |
Cool dry place |
Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Magnesium Stearate, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin | |
Pellet | |
Pellet | |
Rodent |
Safety and Handling
ShelfLife : 1 Year
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.