Learn More
Selleck Chemical LLCSupplier Diversity Partner Dutasteride 100mg 164656-23-9 GI198745, GG-745  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S1202100MG
Dutasteride (GI198745, GG-745) is a dual 5-α reductase inhibitor that inhibits conversion of testosterone to dihydrotestosterone (DHT).
Catalog No. 50-624-8