Learn More
DUX3 Mouse anti-Human, Clone: 3D2, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA524459
Catalog No. PIMA524459
Add to cart
Description
Description
Peptide Sequence: MALLTALDDT LPEEAQGPGR RMILLSTPSQ SDALRACFER NLYPGIATKE QLAQGIDIPE PRVQIWFQNE RSCQLRQHRR QSRPWPGRRD PQKGR
The human genome contains hundreds of repeats of the 3.3-kb family in regions associated with heterochromatin. The DUX gene family, including DUX3, resides within these 3.3-kb repeated elements (Beckers et al., 2001 [PubMed 11245978]). See DUX4 (MIM606009).
Specifications
Specifications
DUX3
Monoclonal
Unconjugated
DUX3
Liquid
DUX3
DUX3 (NP_036280, 1 a.a. ∽ 95 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Protein A
RUO
Antibody
Monoclonal
Human
ELISA, Western Blot
3D2
PBS with no preservative; pH 7.4
Q96PT4
Putative double homeobox protein 3, DUX3
Mouse
IgG2b, kappa
100 μg
-20° C
Primary
26582