Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUX3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548932

Catalog No. PA548932


Add to cart

Description

Description

The human genome contains hundreds of repeats of the 3.3-kb family in regions associated with heterochromatin. The DUX gene family, including DUX3, resides within these 3.3-kb repeated elements (Beckers et al., 2001 [PubMed 11245978]). See DUX4 (MIM606009).
Specifications

Specifications

DUX3
Polyclonal
PBS with 0.09% sodium azide
Q96PT4
Putative double homeobox protein 3, DUX3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
26582
Western Blot
Unconjugated
DUX3
Liquid
DUX3
KLH conjugated synthetic peptide between 21-48 amino acids from the N-terminal region of human DUX3.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit