Learn More
Aviva Systems Biology Corp DUXA Antibody - C-terminal region (ARP47515_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47515_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DUXA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DUXA. Alternate names: -ApplicationWB. Species reactivity: Human
Catalog No. 50-014-59575