Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DVL1 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142878
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
DVL1, the human homolog of the Drosophila dishevelled gene (dsh) encodes a cytoplasmic phosphoprotein that regulates cellproliferation, acting as a transducer molecule for developmentalprocesses, including segmentation and neuroblast specification.DVL1 is a candidate gene for neuroblastomatous transformation. TheSchwartz-Jampel syndrome and Charcot-Marie-Tooth disease type 2Ahave been mapped to the same region as DVL1. The phenotypes ofthese diseases may be consistent with defects which might beexpected from aberrant expression of a DVL gene during development.
Specifications
DVL1 | |
Polyclonal | |
Unconjugated | |
DVL1 | |
dishevelled 1; dishevelled 1 (homologous to Drosophila dsh); dishevelled segment polarity protein 1; dishevelled, dsh homolog 1; dishevelled, dsh homolog 1 (Drosophila); dishevelled-1; Dishevelled-1-like; DRS2; DSH homolog 1; DSH homolog 1-like; Dvl; Dvl1; dvl-1; DVL1L1; DVL1P1; MGC54245; mKIAA4029; RP5-890O3.5; segment polarity protein dishevelled homolog DVL-1 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
13542, 1855 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O14640, P51141 | |
DVL1 | |
Peptide with sequence C-PQLEEAPLTVKSD. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction