Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DVL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA517471
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with Dvl3.
Disheveled DVL2 is a member of the dishevelled (dsh) protein family. The vertebrate dsh proteins have approximately 40% amino acid sequence similarity with Drosophila dsh. This gene encodes a 90-kD protein that undergoes posttranslational phosphorylation to form a 95-kD cytoplasmic protein, which may play a role in the signal transduction pathway mediated by multiple Wnt proteins. The mechanisms of dishevelled function in Wnt signaling are likely to be conserved among metazoans.
Specifications
DVL2 | |
Polyclonal | |
Unconjugated | |
DVL2 | |
dishevelled 2; dishevelled 2 (homologous to Drosophila dsh); dishevelled 2, dsh homolog; dishevelled 2, dsh homolog (Drosophila); dishevelled segment polarity protein 2; dishevelled, dsh homolog 2; dishevelled-2; DSH homolog 2; Dvl2; Dvl-2; LOW QUALITY PROTEIN: segment polarity protein dishevelled homolog DVL-2; sb:eu471; Segment polarity protein dishevelled homolog DVL-2; wu:fc05d12; wu:fo71e09; wu:fp54a02; zgc:55372 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100762587, 13543, 1856, 303251 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunocytochemistry | |
201 μg/mL | |
0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
O14641, Q60838 | |
DVL2 | |
Synthetic peptide corresponding to residues surrounding Ser638 of human Dvl2. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Monkey, Hamster, Mink | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction