Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dyclonine Hydrochloride, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
536-43-6
C18H28ClNO2
325.88
KNZADIMHVBBPOA-UHFFFAOYSA-N
hydrogen 1-(4-butoxyphenyl)-3-(piperidin-1-yl)propan-1-one chloride
[H+].[Cl-].CCCCOC1=CC=C(C=C1)C(=O)CCN1CCCCC1
536-43-6
C18H28ClNO2
325.88
KNZADIMHVBBPOA-UHFFFAOYSA-N
hydrogen 1-(4-butoxyphenyl)-3-(piperidin-1-yl)propan-1-one chloride
[H+].[Cl-].CCCCOC1=CC=C(C=C1)C(=O)CCN1CCCCC1

D1240, 536-43-6, C18H27NO2•HCl

Spectrum™ Dyclonine Hydrochloride, USP is an oral anesthetic. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.002
Melting Range 173-178°C
Loss on Drying 0.01
Quantity 25 g
CAS Max % 1
UN Number UN2811
Percent Purity 98 to 102%