Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DYDC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558107
Description
Immunogen sequence: ELEMQEKERQ RIQELQRAQE QLGKEMRMNM ENLVRNEDIL HSEEATLDSG KTLAEISDRY GAPNLSRVEE LDEPMFSDI Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 62%.
This gene encodes a member of a family of proteins that contains a DPY30 domain. The encoded protein is involved in acrosome formation during spermatid development. This gene locus overlaps with a closely related gene on the opposite strand. Alternative splicing results in multiple transcript variants.
Specifications
DYDC1 | |
Polyclonal | |
Unconjugated | |
DYDC1 | |
1700029M23Rik; bA36D19.5; BC019250 protein; DPY30 domain containing 1; DPY30 domain-containing protein 1; DPY30D1; DYDC1; FLJ43920; OTTHUMP00000019963; RSD9; RSD-9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
143241 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WWB3 | |
DYDC1 | |
Recombinant protein corresponding to Human DYDC1. Recombinant protein control fragment (Product #RP-97167). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction