Learn More
DYKDDDDK Epitope Tag Mouse anti-Nonspecies specific, Alexa Fluor 594, Clone: FG4R, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP237823AF594
Catalog No. NP237823A94
Add to cart
Description
Description
DYKDDDDK Epitope Tag Monoclonal antibody specifically detects DYKDDDDK Epitope Tag in Nonspecies specific samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
DYKDDDDK Epitope Tag
FG4R
Western Blot, Immunocytochemistry/Immunofluorescence
DDDDK epitope tag, DDK, DYKDDDDK tag, ECS epitope tag, ECS tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, epitope tag flag, FLAG (FLAG is a trademark of Sigma Aldrich), Flag tag, Ty 1 Tag, xxxDDDDK epitope tag
DYKDDDDK Epitope Tag Antibody (FG4R) was made to a Synthetic peptide (DYKDDDDK) coupled to KLH.
1.01 kDa
Store at 4°C in the dark.
Monoclonal
Nonspecies specific
Western Blot, Immunocytochemistry, Immunofluorescence
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
Mouse
IgG2b
0.1 mL
Primary
DYKDDDDK Epitope Tag Antibody (FG4R) detects Flag tag from various samples.