Learn More
DYKDDDDK Epitope Tag Mouse anti-Human, Non-species specific, Clone: C.15200054, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259146
Catalog No. NBP259146
Add to cart
Description
Description
DYKDDDDK Epitope Tag Monoclonal antibody specifically detects DYKDDDDK Epitope Tag in Human, Non-species specific samples. It is validated for Western Blot, Chromatin Immunoprecipitation.
Specifications
Specifications
DYKDDDDK Epitope Tag
C.15200054
Western Blot 1:1000, Chromatin Immunoprecipitation 2 ug
Purified
Mouse
IgG1
50 ug
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Monoclonal
Western Blot, ChIP assay
Unconjugated
PBS with 0.05% Sodium Azide
DDDDK epitope tag, DDK, DYKDDDDK epitope tag, DYKDDDDK tag, ECS epitope tag, ECS tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, epitope tag flag, FLAG (FLAG is a trademark of Sigma Aldrich), Flag tag, Ty 1 Tag, xxxDDDDK epitope tag
The exact immunogen is propietary information.
Protein A purified
RUO
Primary
Human, Non-species specific