Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYKDDDDK Tag Mouse, FITC, Clone: 5A8E5, THE™, GenScript™

Catalog No. 502120553
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-212-0553 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-212-0553 Supplier Genscript Corporation Supplier No. A01632
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute the lyophilized product with deionized water (or equivalent) to make antibody concentration of 0.5 mg/mL. GenScript THE™ DYKDDDDK Tag Antibody [FITC], mAb, Mouse is FITC conjugated THE™ DYKDDDDK Tag Antibody, mAb, Mouse (A000187) purified from mice ascites by Protein A affinity column. This product is suitable for detecting the expression level of DYKDDDDK fusion proteins by various immunoassays. Absorption Wavelength: 492 nmEmission Wavelength: 518 nmFluorochrome/Protein Ratio: >3 moles FITC per mole antibody

DYKDDDDK Tag Monoclonal antibody specifically detects DYKDDDDK Tag in Tag samples. It is validated for Flow Cytometry, Immunocytochemistry

Specifications

Antigen DYKDDDDK Tag
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 5A8E5
Conjugate FITC
Formulation PBS with 1% BSA and 0.02% sodium azide; pH 7.4
Gene Alias DDDDK; DDDDK epitope tag; ECS epitope tag; ECS tag; FLAG; FLAG Tag
Host Species Mouse
Immunogen A synthetic peptide DYKDDDDK coupled to KLH
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG2b κ
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.