Learn More
Thermo Scientific™ DyLight™ 405 NHS Ester
Label antibodies or other proteins as probes for fluorescence imaging assays with the full spectral range of NHS ester-activated DyLight Dyes.
$288.00 - $362.00
Products 2
Catalog Number Mfr. No. Description Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Description Quantity Price Quantity & Availability  
PI46400
View Documents
Thermo Scientific™
46400
DyLight 405 NHS Ester 1mg
Each for $362.00
Add to cart
PI46401
View Documents
Thermo Scientific™
46401
DyLight 405 NHS Ester 5 x 50µg
Each for $288.00
Add to cart
Description
Description
Thermo Scientific™ Amine-Reactive DyLight Dyes are NHS ester-activated DyLight Fluors for fluorescent labeling of antibodies and other proteins to be used as molecular probes for cellular imaging and other fluorescence detection methods.
The DyLight Dyes exhibit higher fluorescence intensity and photostability than Alexa™, CyDye™ and LI-COR™ Dyes in many applications and remain highly fluorescent over a broad pH range (pH 4-9). Additionally, the water solubility of the DyLight Dyes allows a high dye-to-protein ratio without precipitation during conjugation.
Highlights:
  • Brightness: high-fluorescence intensity provides outstanding sensitivity and requires less conjugate
  • Photostability: exceptional resistance to photobleaching enables fluorescence imaging under the most demanding conditions (e.g., structured illumination, 4pi microscopy)
  • Instrument compatibility: emission spectra match the principal output wavelengths of common fluorescence instrumentation