Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DyLight™ Far Red Specialty Dyes

Label amine-containing molecules for fluorescence imaging and flow cytometry using these benzopyrillium-based, NHS ester, far red-emitting (647 laser) fluorescent dyes.

Specifications

Quantity 1mg
Sufficient For 1 use to label a total of 7mg of IgG using typical conditions
View More Specs

Products 4
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
PI46635
View Documents
Thermo Scientific™
46635
DyLight 655-B1 NHS Ester This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
PI46636
View Documents
Thermo Scientific™
46636
DyLight 655-B2 NHS Ester This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
PI46637
View Documents
Thermo Scientific™
46637
DyLight 655-B3 NHS Ester This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
PI46638
View Documents
Thermo Scientific™
46638
DyLight 655-B4 NHS Ester This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Thermo Scientific™ DyLight™ Far Red Specialty Dyes are optimized for labeling molecules and other staining applications.

DyLight Far Red-emitting Dyes are a family of labeling agents that provide bright fluorescent detection for imaging. Dyes can be selected based upon their characteristic excitation and emission properties or relative hydrophilicity/hydrophobicity attributes. Dyes that contain a greater number of negatively charged sulfonates generally will have greater water solubility than dyes with fewer sulfonates. More hydrophobic dyes often provide better cell penetrating ability in vivo, while more hydrophilic dyes have less nonspecific binding potential. Each dye contains an amine-reactive NHS ester for simple modification of antibodies, proteins, peptides or other biomolecules through amide bond formation.

Highlights:

  • Large selection–the largest family of dyes available for far red-emitting fluorescence applications

  • NHS ester reactive group–allows immediate labeling of antibodies, proteins, peptides, and other amine-containing molecules through amide bond formation

  • Multiple solubility options –choose from hydrophilic to hydrophobic dyes to optimize the right dye label for the best performance in a given application

Criteria to consider when choosing a DyLight Far Red-emitting Specialty Dye:

  • Excitation and emission wavelengths — choose the best dye to match the excitation and emission capabilities of your instrument
  • Water solubility — choose a dye based on its relative hydrophilicity, which directly correlates to the number of negatively-charged sulfonates it has on its core structure. More hydrophilic dyes are best at maintaining water solubility of a labeled antibody and limiting the nonspecific binding of the conjugate. More hydrophobic dyes often are best at penetrating tissues and cell membranes in vivo, meaning that dyes with fewer sulfonates may work best for some applications.
  • DyLight dye selection — the broad selection of red-emitting dyes allows a number of candidate dyes to be tested in a given application for optimal performance

Recommended for:

Fluorescence imaging; Confocal microscopy; Flow cytometry; Spectral fluorescence imaging; In vivo imaging; Fluorescent western blotting; Protein microarrays; Antibody labeling; Peptide labeling; Fluorescence correlation spectroscopy; Protein arrays; Single molecule detection; Nanoparticle conjugation; Biotin/streptavidin conjugation

Specifications

Specifications

1mg
1 use to label a total of 7mg of IgG using typical conditions
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit