Learn More
Thermo Scientific™ DyLight™ Fluor Antibody Labeling Kits
Label and purify antibodies for fluorescence imaging assays in one hour with the full spectral range of activated DyLight Fluorescent Dye kits.
Specifications
Kit Contents Borate Buffer (0.67M), 1mL
View More Specs
Products 1
Catalog Number Mfr. No. Description Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Description Quantity Price Quantity & Availability  
PI62276
View Documents
Thermo Scientific™
62276
DyLight 350 Antibody Labeling Microscale Kit 5 rxn kit This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Thermo Scientific™ DyLight Fluor Antibody Labeling Kits provide the essential reagents and accessories to easily label and purify antibodies and include regular-scale and microscale kits for each specific DyLight Fluorescent Dye.
Two convenient kit formats are available to accommodate varied labeling requirements. The DyLight Antibody Labeling Kits contain all necessary components to perform three separate labeling reactions using 1mg of IgG or similar quantities of other proteins. The DyLight Microscale Antibody Labeling Kits contain all the necessary components to perform five separate labeling reactions using 100µg of IgG. The labeling kits use high-performance spin desalting columns to provide exceptional dye removal and antibody recovery.
Highlights:
  • Fast - label and purify protein in approximately one hour
  • Amine-reactive dyes - label virtually any protein
  • Efficient non-reacted dye removal
  • Minimal sample dilution
  • Spin column format eliminates the need for column preparation, fraction screening and waiting for protein to emerge from column
Item-Specific Features:
  • Standard kits contain sufficient reagents to label and purify 3 x 1mg of IgG or similar quantities of other proteins
  • Microscale kits contain sufficient reagents to label and purify 5 x 0.1mg of IgG or similar quantities of other proteins
Includes:
Antibody Labeling Kits
DyLight NHS Ester, 3 vials; Reaction buffer, 1mL; Spin columns, 6 each; Microcentrifuge collection tubes, 12 each; Purification resin, 5mL
Microscale Kits
DyLight NHS Ester, 5 vials; Reaction buffer, 1mL; Spin columns, 5 each; Microcentrifuge collection tubes, 10 each; Purification resin, 5mL
Specifications
Specifications
Borate Buffer (0.67M), 1mL