Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DyLight™ Yellow Specialty Dyes

Label amine-containing molecules for fluorescence imaging or flow cytometry with these rhodamine-based, yellow-emitting (532 to 546 laser) NHS ester fluorescent dyes.

Specifications

Quantity 1mg
Sufficient For 1 use to label a total of 7mg of IgG using typical conditions
View More Specs

Products 1
Catalog Number Mfr. No. Formulation Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Price Quantity & Availability  
PI46612
View Documents
Thermo Scientific™
46612
NHS ester activated Rhodamine derivative with 0 sulfonate groups This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Thermo Scientific™ DyLight Yellow-emitting Specialty Dyes are optimized for labeling molecules and other staining applications.

DyLight Yellow-emitting Dyes are a family of labeling agents that provide bright fluorescent detection for imaging. Dyes can be selected based upon their characteristic excitation and emission properties or relative hydrophilicity & hydrophobicity attributes. Dyes that contain a greater number of negatively charged sulfonates generally will have greater water solubility than dyes with fewer sulfonates. More hydrophobic dyes often provide better cell penetrating ability in vivo, while more hydrophilic dyes have less nonspecific binding potential. Each dye contains an amine-reactive NHS ester for simple modification of antibodies, proteins, peptides or other biomolecules through amide bond formation.

Highlights:

  • NHS ester reactive group–allows immediate labeling of antibodies, proteins, peptides and other amine-containing molecules through amide bond formation

  • Multiple solubility options –choose from hydrophilic to hydrophobic dyes to optimize the right dye label for the best performance in a given application

Criteria to consider when choosing a DyLight Yellow-emitting Specialty Dye:

  • Excitation and emission wavelengths–choose the best dye to match the excitation and emission capabilities of your instrument
  • Water solubility–choose a dye based on its relative hydrophilicity, which directly correlates to the number of negatively-charged sulfonates it has on its core structure. More hydrophilic dyes are best at maintaining water solubility of a labeled antibody and limiting the nonspecific binding of the conjugate. More hydrophobic dyes often are best at penetrating tissues and cell membranes in vivo, meaning that dyes with fewer sulfonates may work best for some applications.

Recommended for:

Fluorescence imaging; Fluorescence correlation spectroscopy; Flow cytometry; Imaging; Antibody labeling; Direct immunofluorescence staining; ELISA; Western blotting; Protein microarrays; Polymer labeling; Peptide labeling; Phalloidin labeling for actin staining

Specifications

Specifications

1mg
1 use to label a total of 7mg of IgG using typical conditions
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit