Learn More
Dyn-A-Med Weighing Canoes
Resist dilute acids, aqueous solutions, alcohols and bases
Supplier: Dyn-A-Med 80056
Description
- Anti-static
- Polystyrene
- Contoured sides
Specifications
Polystyrene | |
Ideal for weighing static-affected materials, either liquid or dry | |
7.3 cm | |
3.18 cm |
Weighing Dish Canoe | |
2.88 x 4.75 x 1.25 in. (7.3 x 12.1 x 3.18 cm) | |
12.1 cm |