Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dynabeads™ M-280 Streptavidin Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.

For solation and handling of biotinylated nucleic acids, antibodies, or other biotinylated ligands and targets

Manufacturer:  Invitrogen™ 11206D

 View more versions of this product

Catalog No. 11-206-D


Add to cart

Description

Description

Dynabeads are used on more than 25,000 routine IVD instruments worldwide and are known for exceptional reproducibility (both within and between batches) and automation ability. The strong binding affinity of streptavidin-biotin interaction (Kd=10-15) is used in vast number of applications.

  • Direct and fast isolation of any biotinylated molecule
  • Low-charged and neutral beads, optimal for binding of DNA fragments, proteins, peptides, and antibodies
  • Flexible protocols with gentle and efficient liquid-phase reaction kinetics
  • Biomagnetic protocols are easily adapted to automated platforms
  • High batch-to-batch reproducibility, securingconsistent results in application
  • Bead Diameter: 2.8 µm
  • Concentration: 10mg/mL
  • High throughput compatible
  • Sample Type: Any Sample Type

  • Diameter: 2.8°m, with monolayer, not a multilayer, of recombinant streptavidin covalently coupled to surface and further blocked with BSA
  • Show rapid liquid-phase reaction kinetics.
  • Key applications: Preparing 2 to 5kb samples for mate pair library sequencing, isolation of RNA and DNA binding proteins, immobilization of large DNA fragments, purifying sequencing products, specific capture of nucleic acids
  • Dynabeads™ are used on more than 25,000 routine IVD instruments worldwide
One mg of Dynabeads M-280 Streptavidin typically binds:
  • 650 to 900 pmoles free biotin
  • ~200 pmol biotinylated peptides
  • ~10µg biotinylated IgG
  • ~10µg ds-DNA
  • ~200 pmol ss-oligonucleotides

Antibody Purification, ChIP-on-Chip, Chromatin Biology, Chromatin Immunoprecipitation (ChIP), DNA and RNA Purification and Analysis, DNA Extraction, Immunoprecipitation, Phage Display, Protein and Peptide Purification, Protein Complex Isolation and Analysis, Protein Complex Pull-Down, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis, RNAi, Epigenetics and Non-Coding RNA Research, Sanger Sequencing, Sequencing, Sequencing Reaction Purification

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

2.8 µm
Beads in Suspension
12%
Streptavidin
Research Grade
Tosylactivated, Hydrophobic
10 mg⁄ml, 10 mg/mL
Any Sample Type
This Product Contains: Dynabeads® (2.8 µm) with covalently coupled recombinant streptavidin. Supplied in PBS, pH 7.4 with 0.1% BSA and 0.02% sodium azide.

Store at: 2°C to 8°C.
High-throughput Compatible
pH 5.0
DYNAL®, Dynabeads®
Room Temperature
650-900 pmoles/mg beads
10mL
CV < 3%
Safety and Handling

Safety and Handling

ShelfLife : 36 months from date of manufacture

Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.