Learn More
Invitrogen™ Dynabeads™ SARS-CoV-2 Spike Ig Total ELISA Kit
The Dynabeads™ SARS-CoV-2 Spike Ig Total ELISA Kit is a solid‐phase sandwich enzyme‐linked immunosorbent assay (ELISA) designed to detect and quantify the level of human SARS-CoV-2 IgG, IgM, and IgA in serum and plasma samples. The kit can be used for both qualitative and quantitative assays.
$580.00 - $2470.00
Specifications
Certifications/Compliance | ISO 9001 |
---|---|
Detection Method | Absorbance at 450 nm |
Product Type | SARS-CoV-2 ELISA Ig Total Kit |
For Use With (Application) | Binding of human immunoglobulins (IgG, IgM, IgA) towards SARS-CoV-2 (Spike trimer) |
For Use With (Equipment) | KingFisher™ Flex or Apex Instruments, DynaMag™-96 Side Skirted, Magnetic 96-Well Separator, ELISA Plate Reader (450 nm) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
18020D
![]() |
Invitrogen™
18020D |
96 reactions |
Each for $580.00
|
|
18020DFIVE
![]() |
Invitrogen™
18020DFIVE |
5 x 96 reactions |
Each for $2,470.00
|
|
Description
- A solid‐phase sandwich enzyme‐linked immunosorbent assay (ELISA) designed to detect and quantify the level of human SARS-CoV-2 IgG, IgM, and IgA in serum and plasma samples
- The steps required for use are simply to pre-dilute the reagents, fill the wells of the KingFisher plastic plates, and push start on the instrument
- After the run, the sample is transferred to a plate compatible with the absorbance reader and the OD is read at 450 nm
- A DynaMag-96 well magnet can be used while still taking advantage of the faster binding kinetics of magnetic beads compared to the more traditional plate-bound ELISAs
- Kit contains all buffers required, as well as calibrator antibodies, HRP-conjugated detection antibodies, substrate solution, beads, and stop solution sufficient for one 96-well plate
- Dynabeads SARS-CoV-2 Spike beads can be purchased separately (18100D) for other uses such as the optimization of unique conditions
- Using Dynabeads and automation drastically improves reproducibility, limits hands on time, and reduces operating errors
- The intra-assay CV value is typically <5% (range 2.8–3.2%), while the inter-assay CV is <15% (range 3.9–9.3%)
- Kit can be used for both qualitative and quantitative assays
- One of the fastest ELISAs for SARS-CoV-2 serology
- For research use only, not for use in diagnostic procedures
Specifications
ISO 9001 | |
SARS-CoV-2 ELISA Ig Total Kit | |
KingFisher™ Flex or Apex Instruments, DynaMag™-96 Side Skirted, Magnetic 96-Well Separator, ELISA Plate Reader (450 nm) | |
2.7 μm | |
Yes, on KingFisher™ Flex or Apex instruments |
Absorbance at 450 nm | |
Binding of human immunoglobulins (IgG, IgM, IgA) towards SARS-CoV-2 (Spike trimer) | |
Wet ice (2° to 8°C) | |
Monosized, superparamagnetic polystyrene magnetic beads |
For research use only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.