Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dynactin 4 Mouse anti-Human, FITC, Clone: 3G9D7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237399F

Catalog No. NBP237399F

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

dynactin 4
3G9D7
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
DCTN4
Purified recombinant fragment of human DCTN4 (AA: 57-298) expressed in E. Coli.
Protein A or G purified
Store at 4C in the dark.
Monoclonal
Human
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
FITC
PBS with 0.05% Sodium Azide
dynactin 4 (p62), dynactin p62 subunit, dynactin subunit 4, Dynactin subunit p62
Mouse
IgG1
0.1 ml
Primary
51164
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit