Dynamic Diagnostics Cubitainer™ Carts

Easily elevate and position heavy reagents at instruments

Manufacturer: Dynamic Diagnostics Inc. DDISupplier Diversity Partner 4000106

 View more versions of this product

Catalog No. 22-899-201

  Check Availability
Add to Cart

    Specifications

    Height (English) 5 in.
    Height (Metric) 12.7cm
    Length (English) 26.5 in.
    Length (Metric) 67.3cm
    Width (English) 13.25 in.
    Product Type Cubitainer cart
    Width (Metric) 33.65cm
    Dimensions (L x W x H) 26.5 x 13.25 x 5 in. (67.3 x 33.65 x 12.7cm)
    For Use With (Application) Easily elevate and position heavy reagents at instruments
    Holds Two 20L cubitainer
    Includes 4 Heavy duty 3 in. dia. wheels, 1 in. H side contains spills, built in handles
    Size Double
    Material Powder Coated 22G Steel

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit