Learn More
Dynamic Diagnostics Cubitainer™ Carts
Easily elevate and position heavy reagents at instruments
Manufacturer:  Dynamic Diagnostics Inc. DDISupplier Diversity Partner 4000107
 View more versions of this product
Catalog No. 22-899-202
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Cubitainer cart
5 in.
40 in.
13.25 in.
40 x 13.25 x 5 in. (101.6 x 33.65 x 12.7cm)
Three 20L cubitainer
Triple
Powder Coated 22G Steel
12.7cm
101.6cm
33.65cm
Easily elevate and position heavy reagents at instruments
6 Heavy duty 3 in. dia. Wheels, 1 in. H side contains spills, built-in handles