Learn More
Dynamic Diagnostics Acrylic Safety Splash Shield
Ultimate in flexibility for optimum protection with a 12 × 11 in. curved shield
Manufacturer:  Dynamic Diagnostics 400021
Catalog No. 22-899-019
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
This item is not returnable. View return policy
Description
Description
  • Shield: 11 x 12 in., made from tough shatterproof polycarbonate for maximum optical clarity
  • Resists impact, hot chips, oils, and splashing coolants
  • Curved design maximizes containment of material
  • 18 in. White Gooseneck arm allows for precise positioning
  • Coated arm is easy to clean and keep clean
  • Helps meet compliance with OSHA bloodborne pathogen regulations (29 CFR Part 191.1030)
  • C-clamp mount is padded to protect your desk or benchtop from scratches and can be mounted on a surface up to 2 in. thick
Specifications
Specifications
OSHA bloodborne pathogen regulations (29 CFR Part 191.1030)
Splash shield
Protects you from splashes, aerosol mist, and flying debris
Acrylic