Dynamic Diagnostics Inc. Powder-Coated White Glove Box Holders

22G steel with rust-resistant white powder-coated finish

Manufacturer: Dynamic Diagnostics Inc. DDISupplier Diversity Partner 300010

 View more versions of this product

Catalog No. 22-899-058

  Check Availability
Add to Cart

    Specifications

    Material Steel
    Color White
    For Use With (Application) Side loading design allows easy removal of lower boxes without removing upper boxes
    Includes Mounting hardware
    Style Double
    Product Type Glove Dispenser

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit