Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dynamic Diagnostics TransPak Pre-filled 10% Neutral Buffered Formalin (NBF)
Histology reagent - Pre-filled 10% NBF (Available in different volumes)
Manufacturer: Dynamic Diagnostics MA0102011
Description
- Specimen containers pre-filled with 10% neutral buffered formalin
- Labelled with medical information for clinical use
- Contained in handy dispenser box designed with perforated tear away area on back for easy access
- Available in different volumes and packaging quantity
Specifications
Clear | |
Water, Formaldehyde 37% , Selected Buffers | |
Pre-filled Formalin Container |
Container | |
Primary fixative and followed by secondary fixation by Alcohol, it produces a marvelous set of consistent histologic artifacts and results | |
20mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title