Dynamic Diagnostics TransPak™ Pre-filled 10% Neutral Buffered Formalin (NBF)

Histology reagent - Pre-filled 10% NBF (Available in different volumes)

Manufacturer: Dynamic Diagnostics Inc. DDISupplier Diversity Partner MA0102010FL

 View more versions of this product

Catalog No. 22-899-401

  Check Availability
Add to Cart

    Specifications

    Format Container
    Formulation Water, Formaldehyde 37% , Selected Buffers
    Special Use Primary fixative and followed by secondary fixation by Alcohol, it produces a marvelous set of consistent histologic artifacts and results
    Color Clear
    Product Type Pre-filled Formalin Container
    Quantity 10mL,10mL
    Type Pre-filled Formalin Container

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit