Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynamic Diagnostics TransPak™ Pre-filled 10% Neutral Buffered Formalin (NBF)

Histology reagent - Pre-filled 10% NBF (Available in different volumes)

Manufacturer:  Dynamic Diagnostics Inc. DDISupplier Diversity Partner 0113

 View more versions of this product

Catalog No. 22-899-406


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Bottle
Primary fixative and followed by secondary fixation by Alcohol, it produces a marvelous set of consistent histologic artifacts and results
Pre-filled Formalin Bottle
Water, Formaldehyde 37% , Selected Buffers
Clear
7mL
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit