Dynamic Diagnostics™ Universal Plug Caps

Eliminate the need to purchase size-specific caps

Manufacturer: Dynamic Diagnostics Inc. DDISupplier Diversity Partner 200011DB

 View more versions of this product

Catalog No. 22-899-001

  Check Availability
Add to Cart

    Specifications

    Color Dark Blue
    Product Type Universal Plug Cap
    Closure Type Universal Plug
    For Use With (Equipment) Fits 10, 12, 13 and 16mm tubes
    Size (English) 0.39 to 0.63 in.
    Size (Metric) 10 to 16mm
    Material Polyethylene

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit