Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dynamin 3 Antibody (SR2308), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322364100UL
Description
Dynamin 3 Monoclonal antibody specifically detects Dynamin 3 in Human, Mouse samples. It is validated for Western BlotSpecifications
Dynamin 3 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
SR2308 | |
Western Blot 1:500-1:2000 | |
Dyna III, dynamin 3, dynamin family member, dynamin-3, EC 3.6.5.5, MGC70433, T-dynamin, testicular | |
A synthesized peptide derived from human Dynamin 3 (Uniprot #: Q9UQ16) | |
100 μg | |
Membrane Trafficking and Chaperones | |
26052 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction