Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dynamin 3 Antibody (SR2308), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Dynamin 3 |
---|---|
Clone | SR2308 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
Dynamin 3 Monoclonal antibody specifically detects Dynamin 3 in Human, Mouse samples. It is validated for Western BlotSpecifications
Dynamin 3 | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
Dyna III, dynamin 3, dynamin family member, dynamin-3, EC 3.6.5.5, MGC70433, T-dynamin, testicular | |
A synthesized peptide derived from human Dynamin 3 (Uniprot #: Q9UQ16) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2308 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Membrane Trafficking and Chaperones | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
26052 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title