Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dynamin 3 Polyclonal Antibody
Dynamin 3 Polyclonal Antibody
Supplier: Thermo Scientific PA5116413
Description
Dynamin 3 Polyclonal Antibody for Western Blot, IHC (P), Flow

Specifications
Dynamin 3 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9UQ16 | |
Dnm3 | |
KLH conjugated synthetic peptide between 557-584 amino acids from the Central region of human DNM3. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Dnm3 | |
9630020E24Rik; AW061159; B230343F03Rik; C530045C17; DNM3; Dyn3; Dyna III; dynamin 3; dynamin family member; Dynamin III; dynamin, testicular; dynamin-2; Dynamin-3; KIAA0820; mKIAA0820; T-dynamin; testicular dynamin | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
26052 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction