Learn More
Enzo Life SciencesSupplier Diversity Partner Dynamin monoclonal antibody (D5) (100 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADIVAMSV041E
Clone: D5. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant His6-tagged human dynamin. UniProt: Q05193 (Dynamin-1), P50570 (Dynamin-2), Q9UQ16 (Dynamin-3). Source: Purified from ascites. Species Reactivity: Human, Mouse, Rat, Chicken, Bovine. Applications: Flow Cytometry, IHC, IP, WB. Recommended dilutions: Flow Cytometry (10µg/ml). Immunohistochemistry (1:50, human colon cancer tissue). Western Blot (0.5µg/ml, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.1mM PMSF and 50% glycerol. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-866