Learn More
Thermo Scientific™ DyNAmo ColorFlash Probe qPCR Kit Available on GSA/VA Contract for Federal Government customers only.
Offers equal performance to the DyNAmo Flash Probe qPCR Kit. In addition, it incorporates an innovative multicolor system that ensures correct pipetting.
Manufacturer:  Thermo Scientific™ F456XL
 View more versions of this product
Catalog No. F456XL
Add to cart
Includes: DyNAmo ColorFlash qPCR Mix (2X); 50X ROX dye, 25µM; 40X sample buffer solution with yellow dye
Description
Description
DyNAmo ColorFlash Probe qPCR Kit offers equal performance to the DyNAmo Flash Probe qPCR Kit. In addition, it incorporates an innovative multicolor system that ensures correct pipetting. The 2X master mix contains a blue dye, and a separate sample buffer contains a yellow dye. The qPCR reaction mix containing both components is green. Using this patent-pending multicolor system, pipetting of both the master mix and the sample can be easily monitored. This significantly decreases the risk for pipetting errors during reaction set up, especially when using white reaction vessels. The dyes do not affect the specificity or sensitivity of qPCR assays.
  • Minimized risk of pipetting errors during reaction set up
  • Especially beneficial when using white reaction vessels
  • Extremely fast protocols due to combined annealing and extension step of only 15 sec.
  • Specific and sensitive detection of a wide range of template concentrations
  • dUTP included in the 2X master mix allows the use of UNG for prevention of carry-over contamination
  • This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
DyNAmo™ ColorFlash Probe qPCR Kit
25 x 1mL DyNAmo ColorFlash qPCR Mix (2X); 1 x 1.25mL 50X ROX dye (25μM); 5 x 1mL 40X sample buffer solution with yellow dye
1000 x 50µL Rxns (2500 x 20µL Rxns)
Primer-Probe Detection
DNA, dsDNA
Real time PCR (qPCR), Fast qPCR, qPCR using sequence-specific probes, RT-qPCR using sequence-specific probes
2500 Reactions
Store at -20°C.
5700 Sequence Detection System, 7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT Fast System, 7900HT System, Bio-Rad CFX384 Touch™, BioRad CFX96™, BioRad MyiQ, BioRad iCycler iQ, BioRad iQ5, Cepheid SmartCycler, Corbett Rotor-Gene, Eppendorf MasterCycler RealPlex, MJ Chromo4, MJ Opticon, QuantStudio™, Roche LightCycler 480, StepOne™ System, StepOnePlus™ System, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000, ViiA™ 7 System
Fast
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.