Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DyNamo ColorFlash Probe qPCR Kit

Offers equal performance to the DyNAmo Flash Probe qPCR Kit. In addition, it incorporates an innovative multicolor system that ensures correct pipetting.

Manufacturer:  Thermo Scientific™ F456L

 View more versions of this product

Catalog No. F456L


Add to cart

Includes: DyNAmo ColorFlash qPCR Mix (2X); 50X ROX dye, 25µM; 40X sample buffer solution with yellow dye

Description

Description

DyNAmo ColorFlash Probe qPCR Kit offers equal performance to the DyNAmo Flash Probe qPCR Kit. In addition, it incorporates an innovative multicolor system that ensures correct pipetting. The 2X master mix contains a blue dye, and a separate sample buffer contains a yellow dye. The qPCR reaction mix containing both components is green. Using this patent-pending multicolor system, pipetting of both the master mix and the sample can be easily monitored. This significantly decreases the risk for pipetting errors during reaction set up, especially when using white reaction vessels. The dyes do not affect the specificity or sensitivity of qPCR assays.

  • Minimized risk of pipetting errors during reaction set up
  • Especially beneficial when using white reaction vessels
  • Extremely fast protocols due to combined annealing and extension step of only 15 sec.
  • Specific and sensitive detection of a wide range of template concentrations
  • dUTP included in the 2X master mix allows the use of UNG for prevention of carry-over contamination
Specifications

Specifications

Contains:
• 5 x 1 mL DyNAmo ColorFlash qPCR Mix (2X)
• 1 x 250 µL 50X ROX dye, 25µM
• 1 x 1000 µL 40X sample buffer solution with yellow dye

Store at -20°C
DyNAmo Flash
Primer-Probe Detection
Fast
7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT Fast System, 7900HT System, BioRad CFX96™, QuantStudio™, ViiA™ 7 System, Bio-Rad CFX384 Touch™, Roche LightCycler 480, 5700 Sequence Detection System, StepOnePlus™ System, Eppendorf MasterCycler RealPlex, Corbett RotorGene, Stratagene Mx4000, StepOne™ System, Stratagene Mx3000P, MJ Opticon, MJ Chromo4, BioRad MyiQ, BioRad iQ5, BioRad iCycler iQ, Cepheid SmartCycler, Stratagene Mx3005P
DNA, dsDNA
qPCR
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.