Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DyNAmo Flash Probe qPCR Kit Available on GSA/VA Contract for Federal Government customers only.

Perfect choice for fast probe-based qPCR. The optimized reagents and extremely short cycling times deliver maximum speed thus increasing sample throughput.

Manufacturer:  Thermo Scientific™ F455L

 View more versions of this product

Catalog No. F455L


Add to cart

Includes: DyNAmo Flash qPCR Mix (2X); 50X ROX dye, 25µM

Description

Description

DyNAmo Flash Probe qPCR Kit is the perfect choice for fast probe-based qPCR. The optimized reagents and extremely short cycling times deliver maximum speed thus increasing sample throughput. Significant time savings are achieved with both conventional and fast real-time PCR instruments. Performance of the DyNAmo Flash Probe qPCR Kit is based on an efficient hot start Thermus brockianus DNA polymerase. Sensitivity and the excellent amplification efficiency across a wide range of template concentrations guarantee reproducible results.

DyNAmo Flash Probe qPCR Kit is optimized for hydrolysis probes (e.g., TaqMan probes) and it can be used with both block- and capillary-based realtime PCR instruments.

  • Extremely fast protocols: combined annealing and extension step of only 15 sec.
  • Specific and sensitive detection of a wide range of template concentrations
  • Included dUTP allows the use of UNG for prevention of carry-over contamination
  • Convenient 2X master mix and optimized protocols

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

DyNAmo™ Flash Probe qPCR Kit
5 x 1mL DyNAmo Flash qPCR Mix (2X); 1 x 250μL 50X ROX dye, 25μM
200 x 50µL Rxns (500 x 20µL Rxns)
Primer-Probe Detection
DNA, dsDNA
Real time PCR (qPCR), Fast qPCR, qPCR using sequence-specific probes, RT-qPCR using sequence-specific probes
500 Reactions
Store at -20°C.
5700 Sequence Detection System, 7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT Fast System, 7900HT System, Bio-Rad CFX384 Touch™, BioRad CFX96™, BioRad MyiQ, BioRad iCycler iQ, BioRad iQ5, Cepheid SmartCycler, Corbett Rotor-Gene, Eppendorf™ MasterCycler™ RealPlex, MJ Chromo4, MJ Opticon, QuantStudio™, Roche LightCycler 480, StepOne™ System, StepOnePlus™ System, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000, ViiA™ 7 System
Fast
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.