Learn More
Thermo Scientific™ DyNAmo Flash Probe qPCR Kit
Perfect choice for fast probe-based qPCR. The optimized reagents and extremely short cycling times deliver maximum speed thus increasing sample throughput.
Specifications
Concentration | 2X (DyNAmo Flash qPCR Mix), 50X (ROX Dye) |
---|---|
Detection Method | Primer-Probe |
Format | Tube |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
F455L
|
Thermo Scientific™
F455L |
500 Reactions | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
F455S
|
Thermo Scientific™
F455S |
100 Reactions | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
F455XL
|
Thermo Scientific™
F455XL |
2500 Reactions | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
DyNAmo Flash Probe qPCR Kit is the perfect choice for fast probe-based qPCR. The optimized reagents and extremely short cycling times deliver maximum speed thus increasing sample throughput. Significant time savings are achieved with both conventional and fast real-time PCR instruments. Performance of the DyNAmo Flash Probe qPCR Kit is based on an efficient hot start Thermus brockianus DNA polymerase. Sensitivity and the excellent amplification efficiency across a wide range of template concentrations guarantee reproducible results.
DyNAmo Flash Probe qPCR Kit is optimized for hydrolysis probes (e.g., TaqMan probes) and it can be used with both block- and capillary-based realtime PCR instruments.
- Extremely fast protocols: combined annealing and extension step of only 15 sec.
- Specific and sensitive detection of a wide range of template concentrations
- Included dUTP allows the use of UNG for prevention of carry-over contamination
- Convenient 2X master mix and optimized protocols
Specifications
2X (DyNAmo Flash qPCR Mix), 50X (ROX Dye) | |
Tube | |
qPCR | |
Fast | |
DyNAmo Flash Probe qPCR Kit | |
Real Time PCR SYBR Master Mix |
Primer-Probe | |
High | |
Tbr DNA polymerase | |
DNA, dsDNA | |
DyNAmo Flash | |
Gene Expression |
For Research Use Only. Not for use in diagnostic procedures.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.