Learn More
Thermo Scientific™ DyNAmo HS SYBR Green qPCR Kit
Contains a modified Thermus brockianus DNA polymerase and all the other reagents needed for qPCR.
Supplier: Thermo Scientific™ F410XL
Description
Only template DNA and PCR primers need to be added by the user. The modified DNA polymerase in this kit incorporates a non-specific DNA-binding domain that lends physical stability to the polymerase-DNA complex. ROX passive reference dye is also supplied with the kit in a separate tube for instruments that require ROX normalization.
- Strong specificity: DyNAmo HS SYBR Green master mix minimizes the formation of unwanted primer-dimers and extension of non-specifically bound primers during reaction setup
- Highly sensitive detection: Optimized master mix allows reproducible detection of low-copy number templates
- Broad dynamic range: DyNAmo HS SYBR Green qPCR Kit detects as few as one copy of the target gene and achieves a dynamic range of up to eight orders of magnitude
- Convenient to use: 2X master mix, one tube only; ROX passive reference dye provided in a separate vial
Specifications
Contains: • 25 x1 mL DyNAmo HS SYBR Green qPCR Mix (2X) • 1 x 1.25 mL 50 x ROX dye, 25μM Store at -20°C |
|
Tube | |
qPCR | |
Standard | |
DyNAmo HS SYBR Green qPCR Kit | |
Real Time PCR SYBR Master Mix | |
Gene Expression | |
2500 Reactions | |
25 x 1 mL |
SYBR | |
High | |
Tbr DNA polymerase | |
7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT Fast System, 7900HT System, BioRad CFX96™, QuantStudio™, ViiA™ 7 System, Bio-Rad CFX384 Touch™, Roche LightCycler 480, 5700 Sequence Detection System, StepOnePlus™ System, Eppendorf MasterCycler RealPlex, Corbett RotorGene, StepOne™ System, Stratagene Mx4000, Stratagene Mx3005P, Stratagene Mx3000P, MJ Opticon, MJ Chromo4, Cepheid SmartCycler | |
DyNAmo | |
2500 Reactions of 20 μL Each, 1000 Reactions of 50 μL Each | |
2X (DyNAmo HS SYBR Green qPCR Mix), 50X (ROX Dye) | |
DNA, dsDNA |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.