Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DyNAmo SYBR Green 2-Step qRT-PCR Kit

Perform reliable RNA quantification with this 2-step RT-qPCR Kit.

Specifications

For Use With (Application) Profiling gene expression, Quantification of RNA or DNA targets, Microbial detection, Viral load determination
Includes cDNA synthesis:;M-MuLV RNase H+ reverse transcriptase (includes RNase inhibitor) , 2x RT buffer (includes dNTP mix and MgCl2, random hexamers (300 ng/µl), oligo(dT)15 primer (100 ng/µL).;qPCR:;2x master mix (containing hot start version of modified Therm
Storage Requirements Recommended storage temperature -20°C. Stable for six months from the packaging date.
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
F430L
View Documents
Thermo Scientific™
F430L
100 RT Rxns (200 qPCR Rxns) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

DyNAmo SYBR Green 2-Step RT-qPCR Kit includes all the necessary reagents for cDNA synthesis and SYBR Green qPCR. The reverse transcriptase in the kits is M-MuLV RNase H+, which provides higher sensitivity for qPCR than RNase H- reverse transcriptases. The RNase H activity in the RT enzyme facilitates annealing of PCR primers to the cDNA by degrading the RNA template from the RNA-cDNA hybrid before the PCR step. The performance of the qPCR step is based on a hot start version of a modified Thermus brockianus (Tbr) DNA polymerase.

Highlights

  • Broad dynamic range: up to 1 µg of RNA
  • Reliable results with different RNA sources: mRNA, total RNA, viral RNA and in vitro transcribed RNA
  • Flexible: two-step system, all priming options
  • Optimized 2x mixes for both cDNA synthesis and qPCR
  • Specific and sensitive: uniquely engineered Tbr DNA polymerase
  • Convenient to use: 2x mixes and short protocols
Specifications

Specifications

Profiling gene expression, Quantification of RNA or DNA targets, Microbial detection, Viral load determination
cDNA synthesis:;M-MuLV RNase H+ reverse transcriptase (includes RNase inhibitor) , 2x RT buffer (includes dNTP mix and MgCl2, random hexamers (300 ng/µl), oligo(dT)15 primer (100 ng/µL).;qPCR:;2x master mix (containing hot start version of modified Therm
Recommended storage temperature -20°C. Stable for six months from the packaging date.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit