Learn More
Thermo Scientific™ DyNAzyme EXT DNA Polymerase
Amplify templates up to 40 kb, difficult templates, and GC-rich templates with this optimized mixture of DyNAzyme II DNA Polymerase and a proofreading enzyme.
Manufacturer:  Thermo Scientific™ F505L
 View more versions of this product
Catalog No. F505L
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
DyNAzyme EXT DNA Polymerase is an optimized mixture of DyNAzyme II DNA Polymerase and a proofreading enzyme, capable of amplifying templates up to 40 kb in long PCR reactions.
It is an optimized mixture of DyNAzyme II DNA Polymerase and a proofreading PCR enzyme. DyNAzyme EXT DNA Polymerase offers exceptional performance that Taq-based mixtures cannot match.
  • Ideal for difficult templates, such as GC rich and looped sequences
  • Improved fidelity compared to standard PCR enzymes
  • Generates fragments up to 40 kb in long PCR reactions
  • High yields, robust reactions with standard templates
  • More thermostable than Taq DNA polymerase
DyNAzyme EXT DNA Polymerase is supplied with 10x Optimized DyNAzyme EXT Buffer, 10x Mg2+-free DyNAzyme EXT Buffer, 50 mM MgCl2 -solution, DMSO. Optimized DyNAzyme EXT Buffer provides 1.5 mM MgCl2 in the final 1x concentration.
Recommended for:
Difficult templates, long-range PCR, DHPLC, microarray, cloning
Specifications
Specifications
200U (1U/µL)
DyNAzyme EXT DNA Polymerase
10x Optimized DyNAzyme EXT Buffer, 10x Mg2+-free DyNAzyme EXT Buffer, 50 mM MgCl2 -solution, DMSO. Optimized DyNAzyme EXT Buffer provides 1.5 mM MgCl2 in the final 1x concentration.
Recommended storage temperature -20°C. Stable for one year from the assay date.
Difficult templates, long-range PCR, DHPLC, microarray, cloning