Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DyNAzyme EXT DNA Polymerase

Amplify templates up to 40 kb, difficult templates, and GC-rich templates with this optimized mixture of DyNAzyme II DNA Polymerase and a proofreading enzyme.

Specifications

Description DyNAzyme EXT DNA Polymerase
For Use With (Application) Difficult templates, long-range PCR, DHPLC, microarray, cloning
Includes 10x Optimized DyNAzyme EXT Buffer, 10x Mg2+-free DyNAzyme EXT Buffer, 50 mM MgCl2 -solution, DMSO. Optimized DyNAzyme EXT Buffer provides 1.5 mM MgCl2 in the final 1x concentration.
Storage Requirements Recommended storage temperature -20°C. Stable for one year from the assay date.
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
F505L
View Documents
Thermo Scientific™
F505L
200U (1U/µL) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

DyNAzyme EXT DNA Polymerase is an optimized mixture of DyNAzyme II DNA Polymerase and a proofreading enzyme, capable of amplifying templates up to 40 kb in long PCR reactions.

Highlights

  • Generates fragments up to 40 kb from low complexity templates
  • Ideal for difficult templates, such as GC- rich or looped sequences
  • Improved fidelity compared to standard PCR enzymes
  • High yields, robust reactions with standard templates
Specifications

Specifications

DyNAzyme EXT DNA Polymerase
Difficult templates, long-range PCR, DHPLC, microarray, cloning
10x Optimized DyNAzyme EXT Buffer, 10x Mg2+-free DyNAzyme EXT Buffer, 50 mM MgCl2 -solution, DMSO. Optimized DyNAzyme EXT Buffer provides 1.5 mM MgCl2 in the final 1x concentration.
Recommended storage temperature -20°C. Stable for one year from the assay date.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit