Learn More
Thermo Scientific™ DyNAzyme I & II DNA Polymerases
Get improved thermal stability at high temperatures with these Taq-like enzymes, Thermo Scientific™ DyNAzyme™ DNA Polymerases.
Manufacturer:  Thermo Scientific™ F503L
 View more versions of this product
Catalog No. F503L
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
DyNAzyme DNA Polymerases are Taq-like enzymes with improved thermal stability at high temperatures. They are ideal for all routine PCR applications, and have improved tolerance against DMSO. DyNAzyme I DNA Polymerase is a native thermostable DNA polymerase from Thermus brockianus. DyNAzyme II DNA Polymerase is an easy-to-use, recombinant thermostable DNA polymerase for standard PCR.
Highlights
  • Improved thermal stability
  • Improved tolerance against DMSO
  • Supplied with optimized buffers or with Mg2+-free buffers and MgCl2 solution
  • Detergent-free buffer also available
Specifications
Specifications
DyNAzyme II DNA Polymerase with Mg2+-free buffer
DyNAzyme II DNA Polymerase, Mg-free DyNAzyme Buffer, MgCl2solution
Recommended storage temperature -20 °C. Stable for one year from the assay date.
PCR
1000U (2U/µL)