Learn More
Thermo Scientific™ DyNAzyme I & II DNA Polymerases
Get improved thermal stability at high temperatures with these Taq-like enzymes, Thermo Scientific™ DyNAzyme™ DNA Polymerases.
Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
F501L
View Documents
Thermo Scientific™
F501L
1000U (2U/µL) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
F501S
View Documents
Thermo Scientific™
F501S
250U (2U/µL) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
F503L
View Documents
Thermo Scientific™
F503L
1000U (2U/µL) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
DyNAzyme DNA Polymerases are Taq-like enzymes with improved thermal stability at high temperatures. They are ideal for all routine PCR applications, and have improved tolerance against DMSO. DyNAzyme I DNA Polymerase is a native thermostable DNA polymerase from Thermus brockianus. DyNAzyme II DNA Polymerase is an easy-to-use, recombinant thermostable DNA polymerase for standard PCR.
Highlights
  • Improved thermal stability
  • Improved tolerance against DMSO
  • Supplied with optimized buffers or with Mg2+-free buffers and MgCl2 solution
  • Detergent-free buffer also available