Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYNC1LI1 Mouse anti-Human, Clone: OTI2B6, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525217

Catalog No. PIMA525217


Add to cart

Description

Description

Acts as one of several non-catalytic accessory components of the cytoplasmic dynein 1 complex that are thought to be involved in linking dynein to cargos and to adapter proteins that regulate dynein function. Cytoplasmic dynein 1 acts as a motor for the intracellular retrograde motility of vesicles and organelles along microtubules. May play a role in binding dynein to membranous organelles or chromosomes. Probably involved in the microtubule-dependent transport of pericentrin. Is required for progress throuh the spindle assembly checkpoint. The phosphorylated form appears to be involved in the selective removal of MAD1L1 and MAD1L2 but not BUB1B from kinetochores.
Specifications

Specifications

DYNC1LI1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y6G9
DNCLI1, LIC1, DLC-A, cytoplasmic dynein 1 light intermediate chain 1, dynein light chain A, dynein light chain-A, dynein light intermediate chain 1, cytosolic, dynein, cytoplasmic, light intermediate polypeptide 1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51143
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI2B6
Unconjugated
DYNC1LI1
Liquid
DYNC1LI1
Full length human recombit protein of DYNC1LI1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit