Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DYNLL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 168111AP150UL
Description
The directed movement of membrane-bounded organelles is an important process for various cellular functions including membrane transport, secretion, and axonal transport. Cytoplasmic dynein, a ubiquitous minus end-directed microtubule-based motor protein, is believed to be responsible for the retrograde transport of membranous organelles from the synapse to the cell body in fast axonal transport. The cytoplasmic dynein complex is composed of two ∽530 kDa heavy chains, and intermediate chains of ∽74 kDa and ∽53-59 kDa.Specifications
DYNLL2 | |
Polyclonal | |
Unconjugated | |
DYNLL2 | |
8 kDa dynein light chain b, Dlc2, DLC8b, DNCL1B, Dynein light chain LC8 type 2, DYNLL2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140734, 140735, 68097 | |
-20°C | |
Liquid |
Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q78P75, Q96FJ2, Q9D0M5 | |
Dynll2 | |
DYNLL2 Fusion Protein Ag10345 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction