Learn More
DYNLRB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 150491AP
Catalog No. 50-172-9932
Add to cart
Description
Description
The DYNLRB1 gene is a member of the roadblock dynein light chain family. The encoded cytoplasmic protein is capable of binding intermediate chain proteins, interacts with transforming growth factor-beta, and has been implicated in the regulation of actin modulating proteins. Upregulation of this gene has been associated with hepatocellular carcinomas, suggesting that this gene may be involved in tumor progression. Alternative splicing results in multiple transcript variants. Pseudogenes of this gene have been defined on chromosomes 12 and 18.
Specifications
Specifications
DYNLRB1
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P62627, P62628, Q9NP97
BITH, Bithoraxoid like protein, BLP, DNCL2A, DNLC2A, Dynein associated protein Km23, DYNLRB1, km23, ROBLD1
Rabbit
IgG
150 μL
-20°C
Primary
170714, 67068, 83658
Western Blot
Unconjugated
DYNLRB1
Liquid
Dynlrb1
DYNLRB1 Fusion Protein Ag7032
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat